Anti-MLANA/MART1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-0051R-A750
BOSSBS-0051R-A750EA 890 EUR
BOSSBS-0051R-A750
Anti-MLANA/MART1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Involved in melanosome biogenesis by ensuring the stability of GPR143. Plays a vital role in the expression, stability, trafficking, and processing of melanocyte protein PMEL, which is critical to the formation of stage II melanosomes.

Type: Primary
Antigen: Melanoma antigen recognized by T-cells 1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-118/118
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR