Anti-POU2F2 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-1077R
BOSSBS-1077REA 746 EUR
BOSSBS-1077R
Anti-POU2F2 Rabbit Polyclonal Antibody
Antikörper
Transcription factor that specifically binds to the octamer motif (5'-ATTTGCAT-3'). Regulates transcription in a number of tissues in addition to activating immunoglobulin gene expression. Modulates transcription transactivation by NR3C1, AR and PGR. Isoform OCT2.5 activates the U2 small nuclear RNA (snRNA) promoter. Isoforms OCT2.1, OCT2.2 and OCT2.3 activate octamer-containing promoters. Isoforms OCT2.4 and OCT2.5 repress some promoters and activate others. Isoform OCT2.7 is unable to bind to the octamer motif, but can still activate the beta-casein gene promoter at low levels.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: POU2F2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR