Anti-2-Oct Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-1077R-A680
BOSSBS-1077R-A680EA 899 EUR
BOSSBS-1077R-A680
Anti-2-Oct Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Transcription factor that specifically binds to the octamer motif (5'-ATTTGCAT-3'). Regulates transcription in a number of tissues in addition to activating immunoglobulin gene expression. Modulates transcription transactivation by NR3C1, AR and PGR. Isoform OCT2.5 activates the U2 small nuclear RNA (snRNA) promoter. Isoforms OCT2.1, OCT2.2 and OCT2.3 activate octamer-containing promoters. Isoforms OCT2.4 and OCT2.5 repress some promoters and activate others. Isoform OCT2.7 is unable to bind to the octamer motif, but can still activate the beta-casein gene promoter at low levels.

Type: Primary
Antigen: 2-Oct
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 11-100/463
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR