Anti-OLFM3 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-11061R
BOSSBS-11061REA 719 EUR
BOSSBS-11061R
Anti-OLFM3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: OLFM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR