Anti-IGFBP2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-1108R-A750
BOSSBS-1108R-A750EA 890 EUR
BOSSBS-1108R-A750
Anti-IGFBP2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Inhibits IGF-mediated growth and developmental rates (By similarity). IGF-binding proteins prolong the half-life of the IGFs and have been shown to either inhibit or stimulate the growth promoting effects of the IGFs on cell culture. They alter the interaction of IGFs with their cell surface receptors.

Type: Primary
Antigen: IGFBP2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/305
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR