Anti-AGEs Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-1158R-A680
BOSSBS-1158R-A680EA 899 EUR
BOSSBS-1158R-A680
Anti-AGEs Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Advanced Glycation End products (AGEs) are the result of a chain of chemical reactions after an initial glycation reaction. The intermediate products are known, variously, as Amadori, Schiff base and Maillard products, named after the researchers who first described them. (The litreature is inconsistent in applying these terms. For example, Maillard reaction products are sometimes considered intermediates and sometimes end products.) Side products generated in intermediate steps may be oxidizing agents (such as hydrogen peroxide), or not (such as beta amyloid proteins). 'Glycosylation' is sometimes used for 'glycation' in the litreature, usually as 'non-enzymatic glycosylation. The AGE-modified BSA was produced by reacting BSA with glycolaldehyde under sterile conditions followed by extensive dialysis and purification steps.

Type: Primary
Antigen: AGEs
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR