Anti-GPM6B Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-11847R
BOSSBS-11847REA 719 EUR
BOSSBS-11847R
Anti-GPM6B Rabbit Polyclonal Antibody
Antikörper
GPM6B belongs to the myelin proteolipid protein family and is implicated in neural development. It is expressed in neurons and glia; cerebellar Bergmann glia, in glia within white matter tracts of the cerebellum and cerebrum, and in embryonic dorsal root ganglia.
Recommended Dilutions: Western Blot: 1:100 to 1000; IHC-P: 1:100 to 500; IF(IHC-P): 1:50 to 200.
Type: Primary
Antigen: GPM6B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat