Anti-P2RX7 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-12106R-A680
BOSSBS-12106R-A680EA 874 EUR
BOSSBS-12106R-A680
Anti-P2RX7 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Receptor for ATP that acts as a ligand-gated ion channel. Responsible for ATP-dependent lysis of macrophages through the formation of membrane pores permeable to large molecules. Could function in both fast synaptic transmission and the ATP-mediated lysis of antigen-presenting cells.

Type: Primary
Antigen: P2RX7
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 115-160/595
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR