Anti-P2RX7 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-12106R-A750
BOSSBS-12106R-A750EA 874 EUR
BOSSBS-12106R-A750
Anti-P2RX7 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Receptor for ATP that acts as a ligand-gated ion channel. Responsible for ATP-dependent lysis of macrophages through the formation of membrane pores permeable to large molecules. Could function in both fast synaptic transmission and the ATP-mediated lysis of antigen-presenting cells.
Type: Primary
Antigen: P2RX7
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 115-160/595
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse