Anti-P2RX7 Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-12106R-CY7
BOSSBS-12106R-CY7EA 876 EUR
BOSSBS-12106R-CY7
Anti-P2RX7 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Receptor for ATP that acts as a ligand-gated ion channel. Responsible for ATP-dependent lysis of macrophages through the formation of membrane pores permeable to large molecules. Could function in both fast synaptic transmission and the ATP-mediated lysis of antigen-presenting cells.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: P2RX7
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat