Anti-ZNF81 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-12210R-A680
BOSSBS-12210R-A680EA 890 EUR
BOSSBS-12210R-A680
Anti-ZNF81 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kr_ppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. ZNF81, also known as HFZ20 or MRX45, is a transcriptional regulator belonging to the Kr_ppel C2H2-type zinc-finger protein family. It localizes to the nucleus and contains 12 C2H2-type zinc fingers and 1 KRAB domain. Mutations in the gene encoding ZNF81 are implicated in nonsyndromic X-linked mental retardation (XLMR).

Type: Primary
Antigen: ZNF81
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 601-661/661
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR