Anti-ZNF342/ZNF296 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-12212R-A750
BOSSBS-12212R-A750EA 890 EUR
BOSSBS-12212R-A750
Anti-ZNF342/ZNF296 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kr_ppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. As a member of the Kr_ppel C2H2-type zinc-finger protein family, ZNF342 (zinc finger protein 342), also known as Zinc finger protein 296, is a 475 amino acid nuclear protein that contains six C2H2-type zinc fingers through which it is thought to be involved in DNA-binding and transcriptional regulation.

Type: Primary
Antigen: ZNF342/ZNF296
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 231-350/475
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR