Anti-ZNF750 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-12239R-A750
BOSSBS-12239R-A750EA 890 EUR
BOSSBS-12239R-A750
Anti-ZNF750 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Zinc finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc finger proteins contain a Kr_ppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. Zinc finger protein 750 is a 723 amino acid member of the Kr_ppel C2H2-type zinc finger protein family. localised to the nucleus, ZNF750 contains one conserved C2H2 zinc finger domain and is expressed in the skin, lungs, prostate, placenta and thymus. ZNF750 is also expressed in primary human keratinocytes but not in fibroblasts. Mutations in the gene encoding ZNF750 cause Seborrhea-like dermatitis with psoriasiform, a condition characterised by a chronic and diffuse rash on the face and hyperkeratosis of skin over the elbows, soles, knees and palms.

Type: Primary
Antigen: ZNF750
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/723
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR