Anti-Annexin A13 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-12478R-A680
BOSSBS-12478R-A680EA 890 EUR
BOSSBS-12478R-A680
Anti-Annexin A13 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
The Annexins constitute a family of structurally-related, relatively abundant proteins that exhibit Ca²⁺ dependent binding to phospholipids. Annexins function in multiple aspects of cell biology including regulation of membrane trafficking, transmembrane channel activity, inhibition of phospholipase A2, inhibition of coagulation and mediation of cell-matrix interactions. Annexin A13 is considered the original progenitor of the 12 members of vertebrate Annexins. The expression of Annexin A13 is highly tissue-specific, being expressed only in intestinal and kidney epithelial cells. This expression is associated with a highly differentiated intracellular transport function. Two alternative splicing isoforms of Annexin A13 exist, both of which bind to rafts.
Type: Primary
Antigen: Annexin A13
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 241-316/316
Host: Rabbit
Isotype: IgG
Reactivity: Human