Anti-ANXA13 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-12478R-CY7
BOSSBS-12478R-CY7EA 893 EUR
BOSSBS-12478R-CY7
Anti-ANXA13 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
The Annexins constitute a family of structurally-related, relatively abundant proteins that exhibit Ca2+-dependent binding to phospholipids. Annexins function in multiple aspects of cell biology including regulation of membrane trafficking, transmembrane channel activity, inhibition of phospholipase A2, inhibition of coagulation and mediation of cell-matrix interactions. Annexin A13 is considered the original progenitor of the 12 members of vertebrate Annexins. The expression of Annexin A13 is highly tissue-specific, being expressed only in intestinal and kidney epithelial cells. This expression is associated with a highly differentiated intracellular transport function. Two alternative splicing isoforms of Annexin A13 exist, both of which bind to rafts.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ANXA13
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR