Anti-DPP3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-13029R-A750
BOSSBS-13029R-A750EA 890 EUR
BOSSBS-13029R-A750
Anti-DPP3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Dipeptidyl peptidases (DPPs) mediate regulatory activity of their substrates and have been linked to a variety of diseases including type 2 diabetes, obesity and cancer. DPPs have post-proline dipeptidyl aminopeptidase activity, cleaving Xaa-Pro dipeptides from the N-termini of proteins. DPPs can bind specific voltage-gated potassium channels and alter their expression and biophysical properties and may also influence T cells. DPP proteins include DPRP1, DPRP2, DPP3, DPP7, DPP10, DPPX and CD26. DPP3 (dipeptidyl-peptidase 3), also known as DPPIII, is a zinc-exopeptidase that belongs to the peptidase M49 family. DPP3 localizes to the cytoplasm and is involved in intracellular protein catabolism. More specifically, DPP3 is an important enzyme involved in the degradation of enkephalins. An increase in the activity of DPP3 is implicated in ovarian and endometrial cancers.

Type: Primary
Antigen: DPP3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/737
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR