Anti-ERGI3/ERGIC-3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-13103R-A680
BOSSBS-13103R-A680EA 890 EUR
BOSSBS-13103R-A680
Anti-ERGI3/ERGIC-3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Cycling proteins play important roles in the organization and function of the early secretory pathway by participating in membrane traffic and selective transport of cargo between the endoplasmic reticulum (ER), the intermediate compartment (ERGIC), and the Golgi. A family of membrane bound, ubiquitous proteins involved in the selective transport of newly synthesized glycoproteins from the ER to the ERGIC include VIP36, ERGIC-53, ERGIC-1, ERGIC-2 and ERGIC-3. ERGIC-1, also designated ERGIC32, is thought to modulate the activity of a complex formed by ERGIC-2, also designated Erv41, and ERGIC-3, also designated Erv46. ERGIC-2 and ERGIC-3 are both mammalian homologs of yeast proteins abundant in COPII-coated vesicles and localize to the Cis-face of the Golgi apparatus.

Type: Primary
Antigen: ERGI3/ERGIC-3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 301-383/383
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR