Anti-ERGIC3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-13103R-HRP
BOSSBS-13103R-HRPEA 893 EUR
BOSSBS-13103R-HRP
Anti-ERGIC3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Cycling proteins play important roles in the organization and function of the early secretory pathway by participating in membrane traffic and selective transport of cargo between the endoplasmic reticulum (ER), the intermediate compartment (ERGIC), and the Golgi. A family of membrane bound, ubiquitous proteins involved in the selective transport of newly synthesized glycoproteins from the ER to the ERGIC include VIP36, ERGIC-53, ERGIC-1, ERGIC-2 and ERGIC-3. ERGIC-1, also designated ERGIC32, is thought to modulate the activity of a complex formed by ERGIC-2, also designated Erv41, and ERGIC-3, also designated Erv46. ERGIC-2 and ERGIC-3 are both mammalian homologs of yeast proteins abundant in COPII-coated vesicles and localize to the Cis-face of the Golgi apparatus.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: ERGIC3
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR