Anti-ETNK2 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-13112R-CY7
BOSSBS-13112R-CY7EA 893 EUR
BOSSBS-13112R-CY7
Anti-ETNK2 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Ethanolamine kinase 2, also known as EKI2, ETNK2 or HMFT1716, is a 386 amino acid protein that belongs to the choline/ethanolamine kinase family. Via the cytidine diphosphate (CDP) ethanolamine pathway, Ethanolamine kinase 2 catalyses the initial step of phosphatidylethanolamine (PtdEtn) biosynthesis. Ethanolamine kinase 2 is expressed in kidney, liver, testis, ovary and prostate, and is highly specific for ethanolamine phosphorylation. Upregulated during testis development, Ethanolamine kinase 2 may play an essential role in regulating placental hemostasis. Existing as three alternatively spliced isoforms, the gene encoding Ethanolamine kinase 2 maps to human and mouse chromosome 1. Human chromosome 1 spans 260 million base pairs, contains over 3,000 genes, comprises nearly 8% of the human genome and houses a large number of disease-associated genes, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson’s disease, Gaucher disease, schizophrenia and Usher syndrome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ETNK2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR