Anti-ZBTB22 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BS-13566R-A555
BOSSBS-13566R-A555EA 893 EUR
BOSSBS-13566R-A555
Anti-ZBTB22 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
ZBTB22 contains 1 BTB (POZ) domain and 3 C2H2-type zinc fingers and belongs to the krueppel C2H2-type zinc finger protein family. ZBTB22 may be involved in transcriptional regulation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ZBTB22
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat