Anti-ZBTB22 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-13566R-HRP
BOSSBS-13566R-HRPEA 893 EUR
BOSSBS-13566R-HRP
Anti-ZBTB22 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
ZBTB22 contains 1 BTB (POZ) domain and 3 C2H2-type zinc fingers and belongs to the krueppel C2H2-type zinc finger protein family. ZBTB22 may be involved in transcriptional regulation.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: ZBTB22
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR