Anti-ZBTB39 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-13573R-CY3
BOSSBS-13573R-CY3EA 893 EUR
BOSSBS-13573R-CY3
Anti-ZBTB39 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
ZBTB39 may be involved in transcriptional regulation. It belongs to the krueppel C2H2-type zinc-finger protein family and contains one BTB (POZ) domain and eight C2H2-type zinc fingers.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZBTB39
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR