Anti-ZBTB5 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-13581R-A750
BOSSBS-13581R-A750EA 890 EUR
BOSSBS-13581R-A750
Anti-ZBTB5 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kr_ppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. Zinc finger and BTB domain-containing protein 5 (ZBTB5) is a 677 amino acid member of the Kr_ppel C2H2-type zinc-finger protein family. localised to the nucleus, ZBTB5 contains a BTB domain, also known as a POZ domain, which inhibits DNA binding and mediates homotypic and heterotypic dimerization. Characteristics of the BTB domain suggest that ZBTB5 functions as a transcription regulator.
Type: Primary
Antigen: ZBTB5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 581-677/677
Host: Rabbit
Isotype: IgG
Reactivity: Human