Anti-ZNF575 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-13588R-A680
BOSSBS-13588R-A680EA 890 EUR
BOSSBS-13588R-A680
Anti-ZNF575 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kr_ppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. Zinc finger protein 575 (ZNF575) is a 245 amino acid member of the Kr_ppel C2H2-type zinc-finger protein family. Localized to the nucleus, ZNF575 contains six C2H2-type zinc fingers through which it is thought to be involved in DNA-binding and transcriptional regulation.
Type: Primary
Antigen: ZNF575
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/245
Host: Rabbit
Isotype: IgG
Reactivity: Rat