Anti-ZNF672 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-13589R-A750
BOSSBS-13589R-A750EA 890 EUR
BOSSBS-13589R-A750
Anti-ZNF672 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
ZNF672 is a 452 amino acid nuclear protein that may be involved in transcriptional regulation. Belonging to the Kr_ppel C2H2-type zinc-finger protein family, ZNF672 contains 14 C2H2-type zinc fingers. ZNF672 exists as two alternatively spliced isoforms, and is encoded by a gene that maps to human chromosome 1q44. Human chromosome 1 spans 260 million base pairs, contains over 3,000 genes, comprises nearly 8% of the human genome, and houses a large number of disease-associated genes, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson?s disease, Gaucher disease, schizophrenia and Usher syndrome.

Type: Primary
Antigen: ZNF672
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-300/452
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR