Anti-ZNF323 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-13604R-A680
BOSSBS-13604R-A680EA 890 EUR
BOSSBS-13604R-A680
Anti-ZNF323 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. As a member of the Kr_ppel C2H2-type zinc-finger protein family, ZNF323 (zinc finger protein 323) is a 406 amino acid protein containing six C2H2-type zinc fingers and one SCAN box domain. Specifically, C2H2-type zinc fingers function to bind DNA, while SCAN box domains are thought to participate in protein-protein interactions. Therefore, it is probable that ZNF323 functions as a transcription factor. With highest expression in kidney, liver and lung and weaker expression in brain, heart, intestine, muscle, cholecyst and pancreas, ZNF323 is localized to the nucleus. It is also suggested that ZNF323 may play a role in the development of multiple embryonic organs.

Type: Primary
Antigen: ZNF323
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 151-250/406
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR