Anti-TMEM115 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-13697R-CY7
BOSSBS-13697R-CY7EA 893 EUR
BOSSBS-13697R-CY7
Anti-TMEM115 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Transmembrane proteins contain transmembrane domains that are usually characterized by alpha-helical structures. Transmembrane proteins exist as thermodynamically stable hetero- and homodimers that interact with the lipid bilayer and are involved in both material exchange and communication between the cell and the environment. PL6 is a 351 amino acid multi-pass membrane protein that is highly expressed in kidney and skeletal muscle with lower levels of expression detected in liver, placenta, prancreas, lung, heart and brain. PL6 contains one phosphoserine residue and several transmembrane domains, suggesting that it may participate in protein exchange and signaling events between cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TMEM115
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR