Anti-YY1 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-1415R
BOSSBS-1415REA 1090 EUR
BOSSBS-1415R
Anti-YY1 Rabbit Polyclonal Antibody
Antikörper
YY1 is a ubiquitously distributed transcription factor belonging to the GLI Kruppel class of zinc finger proteins. The protein is involved in repressing and activating a diverse number of promoters. YY1 may direct histone deacetylases and histone acetyltransferases to a promoter in order to activate or repress the promoter, thus implicating histone modification in the function of YY1.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: YY1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR