Anti-C7ORF25 Rabbit Polyclonal Antibody (ALEXA FLUOR® 555)

Lieferant: Bioss
BS-15260R-A555
BOSSBS-15260R-A555EA 893 EUR
BOSSBS-15260R-A555
Anti-C7ORF25 Rabbit Polyclonal Antibody (ALEXA FLUOR® 555)
Antikörper
Chromosome 7 has been linked to Osteogenesis imperfecta, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome. The deletion of a portion of the q arm of chromosome 7 is associated with Williams-Beuren syndrome, a condition characterised by mild mental retardation, an unusual comfort and friendliness with strangers and an elfin appearance. Deletions of portions of the q arm of chromosome 7 are also seen in a number of myeloid disorders including cases of acute myelogenous leukemia and myelodysplasia. The C7orf25 gene product has been provisionally designated C7orf25 pending further characterisation.

Type: Primary
Antigen: C7orf25
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 555
Public Immunogen Range: 1-100/421
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR