Anti-C7ORF44 Rabbit Polyclonal Antibody (ALEXA FLUOR® 555)
Lieferant: Bioss
BS-15269R-A555
BOSSBS-15269R-A555EA 893 EUR
BOSSBS-15269R-A555
Anti-C7ORF44 Rabbit Polyclonal Antibody (ALEXA FLUOR® 555)
Antikörper
Chromosome 7 has been linked to Osteogenesis imperfecta, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome. The deletion of a portion of the q arm of chromosome 7 is associated with Williams-Beuren syndrome, a condition characterised by mild mental retardation, an unusual comfort and friendliness with strangers and an elfin appearance. Deletions of portions of the q arm of chromosome 7 are also seen in a number of myeloid disorders including cases of acute myelogenous leukemia and myelodysplasia. The C7orf44 gene product has been provisionally designated C7orf44 pending further characterisation.
Type: Primary
Antigen: C7orf44
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 555
Public Immunogen Range: 51-146/146
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen