Anti-C7ORF46 Rabbit Polyclonal Antibody (Cy5)

Lieferant: Bioss
BS-15271R-CY5
BOSSBS-15271R-CY5EA 893 EUR
BOSSBS-15271R-CY5
Anti-C7ORF46 Rabbit Polyclonal Antibody (Cy5)
Antikörper
Chromosome 7 has been linked to Osteogenesis imperfecta, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome. The deletion of a portion of the q arm of chromosome 7 is associated with Williams-Beuren syndrome, a condition characterised by mild mental retardation, an unusual comfort and friendliness with strangers and an elfin appearance. Deletions of portions of the q arm of chromosome 7 are also seen in a number of myeloid disorders including cases of acute myelogenous leukemia and myelodysplasia. The C7orf46 gene product has been provisionally designated C7orf46 pending further characterisation.

Type: Primary
Antigen: C7orf46
Clonality: Polyclonal
Clone:
Conjugation: Cy5
Public Immunogen Range: 201-298/298
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR