Anti-C9ORF66 Rabbit Polyclonal Antibody (FITC)

Lieferant: Bioss
BS-15337R-FITC
BOSSBS-15337R-FITCEA 893 EUR
BOSSBS-15337R-FITC
Anti-C9ORF66 Rabbit Polyclonal Antibody (FITC)
Antikörper
Chromosome 9 consists of about 145 million bases and 4% of the human genome and encodes nearly 900 genes. Considered to play a role in gender determination, deletion of the distal portion of 9p can lead to development of male to female sex reversal, the phenotype of a female with a male X,Y genotype. Hereditary hemorrhagic telangiectasia, which is characterised by harmful vascular defects, is associated with the chromosome 9 gene encoding endoglin protein, ENG. Familial dysautonomia is also associated with chromosome 9 though through the gene IKBKAP. Notably, chromosome 9 encompasses the largest interferon family gene cluster. Chromosome 9 is partnered with chromosome 22 in the translocation leading to the aberrant production of BCR-ABL fusion protein often found in leukemias. The C9orf66 gene product has been provisionally designated C9orf66 pending further characterisation.

Type: Primary
Antigen: C9orf66
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 1-100/295
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR