Anti-NQO1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-2184R-CY3
BOSSBS-2184R-CY3EA 901 EUR
BOSSBS-2184R-CY3
Anti-NQO1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
The enzyme apparently serves as a quinone reductase in connection with conjugation reactions of hydroquinons involved in detoxification pathways as well as in biosynthetic processes such as the vitamin K-dependent gamma-carboxylation of glutamate residues in prothrombin synthesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NQO1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR