Anti-Vimentin Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)

Lieferant: Bioss
BS-23063R-A750
BOSSBS-23063R-A750EA 899 EUR
BOSSBS-23063R-A750
Anti-Vimentin Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)
Antikörper
Vimentins are class-III intermediate filaments found in various non-epithelial cells, especially mesenchymal cells. Vimentin is attached to the nucleus, endoplasmic reticulum, and mitochondria, either laterally or terminally. Involved with LARP6 in the stabilisation of type I collagen mRNAs for CO1A1 and CO1A2.

Type: Primary
Antigen: Vimentin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/466
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR