Anti-PXR Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-2334R-A350
BOSSBS-2334R-A350EA 901 EUR
BOSSBS-2334R-A350
Anti-PXR Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Nuclear receptor that binds and is activated by variety of endogenous and xenobiotic compounds. Transcription factor that activates the transcription of multiple genes involved in the metabolism and secretion of potentially harmful xenobiotics, drugs and endogenous compounds. Activated by the antibiotic rifampicin and various plant metabolites, such as hyperforin, guggulipid, colupulone, and isoflavones. Response to specific ligands is species-specific. Activated by naturally occurring steroids, such as pregnenolone and progesterone. Binds to a response element in the promoters of the CYP3A4 and ABCB1/MDR1 genes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PXR
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR