Anti-CNR2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-2377R-A350
BOSSBS-2377R-A350EA 876 EUR
BOSSBS-2377R-A350
Anti-CNR2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Heterotrimeric G protein-coupled receptor for endocannabinoid 2-arachidonoylglycerol mediating inhibition of adenylate cyclase. May function in inflammatory response, nociceptive transmission and bone homeostasis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CNR2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR