Anti-CNR2/CB2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-2377R-A680
BOSSBS-2377R-A680EA 874 EUR
BOSSBS-2377R-A680
Anti-CNR2/CB2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Heterotrimeric G protein-coupled receptor for endocannabinoid 2-arachidonoylglycerol mediating inhibition of adenylate cyclase. May function in inflammatory response, nociceptive transmission and bone homeostasis.

Type: Primary
Antigen: CNR2/CB2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 251-350/360
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR