Anti-CNR2 Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-2377R-CY7
BOSSBS-2377R-CY7EA 876 EUR
BOSSBS-2377R-CY7
Anti-CNR2 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Heterotrimeric G protein-coupled receptor for endocannabinoid 2-arachidonoylglycerol mediating inhibition of adenylate cyclase. May function in inflammatory response, nociceptive transmission and bone homeostasis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CNR2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen