Anti-ZNF217 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-2510R-A680
BOSSBS-2510R-A680EA 874 EUR
BOSSBS-2510R-A680
Anti-ZNF217 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
ZNF217 (Zinc-finger protein 217) is a Kruppel-like zinc-finger protein which functions as a transcriptional repressor protein and forms a complex with a number of proteins including CoREST, BHC110/LSD1, HDAC2 and CtBP1. ZNF217 has been shown to repress E-cadherin promoter activity. ZNF217 is overexpressed in a number of cancers including breast and ovarian. ZNF217 and its associated proteins have been implicated in a novel pathway that is involved in cancer progression.
Type: Primary
Antigen: ZNF217
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 151-250/1061
Host: Rabbit
Isotype: IgG
Reactivity: Human