Anti-ZNF217 Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BS-2510R-CY5.5
BOSSBS-2510R-CY5.5EA 876 EUR
BOSSBS-2510R-CY5.5
Anti-ZNF217 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
ZNF217 (Zinc-finger protein 217) is a Kruppel-like zinc-finger protein which functions as a transcriptional repressor protein and forms a complex with a number of proteins including CoREST, BHC110/LSD1, HDAC2 and CtBP1. ZNF217 has been shown to repress E-cadherin promoter activity. ZNF217 is overexpressed in a number of cancers including breast and ovarian. ZNF217 and its associated proteins have been implicated in a novel pathway that is involved in cancer progression.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ZNF217
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat