Anti-IBSP Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BS-2668R-A555
BOSSBS-2668R-A555EA 876 EUR
BOSSBS-2668R-A555
Anti-IBSP Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Binds tightly to hydroxyapatite. Appears to form an integral part of the mineralized matrix. Probably important to cell-matrix interaction. Promotes Arg-Gly-Asp-dependent cell attachment.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: IBSP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat