Anti-FANCG Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-4106R-CY3
BOSSBS-4106R-CY3EA 893 EUR
BOSSBS-4106R-CY3
Anti-FANCG Rabbit Polyclonal Antibody (Cy3®)
Antikörper
FANCG, involved in Fanconi anemia, confers resistance to both hygromycin and mitomycin C. FANCG contains a 5-prime GC-rich untranslated region characteristic of housekeeping genes. The putative 622-amino acid protein has a leucine-zipper motif at its N-terminus. Fanconi anemia is an autosomal recessive disorder with diverse clinical symptoms, including developmental anomalies, bone marrow failure, and early occurrence of malignancies. A minimum of 8 FA genes have been identified.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: FANCG
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat