Anti-ZNF431 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-6120R-A647
BOSSBS-6120R-A647EA 893 EUR
BOSSBS-6120R-A647
Anti-ZNF431 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
ZNF431 belongs to the krueppel C2H2-type zinc-finger protein family and is probably involved in transcriptional regulation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZNF431
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR