Anti-NMT2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-6233R-A555
BOSSBS-6233R-A555EA 893 EUR
BOSSBS-6233R-A555
Anti-NMT2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
NMT2 catalyzes the reaction of N-terminal myristoylation of many signaling proteins. It transfers myristic acid from myristoyl coenzyme A to the amino group of a protein's N-terminal glycine residue. several distinct NMTs exist, varying in apparent molecular weight and /or subcellular distribution.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NMT2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR