Anti-GDF5 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-6580R
BOSSBS-6580REA 719 EUR
BOSSBS-6580R
Anti-GDF5 Rabbit Polyclonal Antibody
Antikörper
Defects in GDF5 are the cause of acromesomelic chondrodysplasia Grebe type (AMDG) . Acromesomelic chondrodysplasias are rare hereditary skeletal disorders characterized by short stature, very short limbs, and hand/foot malformations. The severity of limb abnormalities increases from proximal to distal with profoundly affected hands and feet showing brachydactyly and/or rudimentary fingers (knob-like fingers). AMDG is an autosomal recessive form characterized by normal axial skeletons and missing or fused skeletal elements within the hands and feet.Defects in GDF5 are the cause of acromesomelic chondrodysplasia Hunter-Thompson type (AMDH). AMDH is an autosomal recessive form of dwarfism. Patients have limb abnormalities, with the middle and distal segments being most affected and the lower limbs more affected than the upper. AMDH is characterized by normal axial skeletons and missing or fused skeletal elements within the hands and feet.Defects in GDF5 are the cause of brachydactyly type C (BDC). BDC is an autosomal dominant disorder characterized by an abnormal shortness of the fingers and toes.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: CDMP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR