Anti-DDCT Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-6608R
BOSSBS-6608REA 719 EUR
BOSSBS-6608R
Anti-DDCT Rabbit Polyclonal Antibody
Antikörper
Tautomerization of D-dopachrome with decarboxylation to give 5,6-dihydroxyindole (DHI).

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: DDCT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR