Anti-CRHR2 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-6620R
BOSSBS-6620REA 719 EUR
BOSSBS-6620R
Anti-CRHR2 Rabbit Polyclonal Antibody
Antikörper
This is a receptor for corticotropin releasing factor. Shows high-affinity CRF binding. Also binds to urocortin I, II and III. The activity of this receptor is mediated by G proteins which activate adenylyl cyclase.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: CRHR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR