Anti-FAM89B Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-6758R-CY3
BOSSBS-6758R-CY3EA 893 EUR
BOSSBS-6758R-CY3
Anti-FAM89B Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Mtvr1 is a 176 amino acid protein that exists as two alternatively spliced isoforms. Belonging to the FAM89 family, Mtvr1 is encoded by a gene that maps to human chromosome 11, which comprises approximately 4% of human genomic DNA and is considered a gene and disease association dense chromosome. The chromosome 11 encoded Atm gene is important for regulation of cell cycle arrest and apoptosis following double strand DNA breaks. Atm mutation leads to the disorder known as ataxia-telangiectasia. The blood disorders Sickle cell anemia and thalassemia are caused by HBB gene mutations, while Wilms' tumors, WAGR syndrome and Denys-Drash syndrome are associated with mutations of the WT1 gene. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitz syndrome are also associated with defects in chromosome 11-encoded genes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FAM89B
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR