Anti-IFNA21 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-7021R-CY7
BOSSBS-7021R-CY7EA 893 EUR
BOSSBS-7021R-CY7
Anti-IFNA21 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
This gene is a member of the alpha interferon gene cluster on the short arm of chromosome 9. Interferons are cytokines produced in response to viral infection that mediate the immune response and interfere with viral replication. The encoded protein is a type I interferon and may play a specific role in the antiviral response to rubella virus. [provided by RefSeq, Sep 2011].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IFNA21
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR