Anti-NUBP1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss
BS-7817R-FITC
BOSSBS-7817R-FITCEA 893 EUR
BOSSBS-7817R-FITC
Anti-NUBP1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Implicated in the regulation of centrosome duplication (By similarity). Component of the cytosolic iron-sulfur (Fe/S) protein assembly machinery. Required for maturation of extramitochondrial Fe/S proteins. May bind and transfer 2 labile 4Fe-4S clusters to target apoproteins.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NUBP1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR